Learn More
DWK Life Sciences Kimble™ Kontes™ Open-Top Compression Caps for Microscale Chemistry Kits
Description
- Glass-filled nylon cap with modified GPI thread finish
- Accommodate us with an O-ring seal
- Temperature limit: 150°C
Specifications
Specifications
| For Use With (Equipment) | MICROFLEX™ microscale chemistry kits, for 14/10 connections |
| Material | Nylon |
| Thread Size | 20-400 |
| Product Type | Closure |