Learn More
DWK Life Sciences Kimble™ Kontes™ Open-Top Compression Caps for Microscale Chemistry Kits
For use with MICROFLEX™ microscale chemistry kits
Supplier: DWK Life Sciences 4101191307
Description
- Glass-filled nylon cap with modified GPI thread finish
- Accommodate us with an O-ring seal
- Temperature limit: 150°C
Chemical Identifiers
| Nylon |
Specifications
| MICROFLEX™ microscale chemistry kits | |
| Nylon | |
| 13-425 | |
| Compression Cap |