Learn More
DWK Life Sciences Kimble™ Glass Dropping Bottles
Convenient all-in-one configuration
$43.79 - $45.09
Specifications
| Includes | Ground in Glass pipet and rubber bulb |
|---|---|
| Material | Borosilicate Glass |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
02992A
|
DWK Life Sciences
FS1503530 |
30 mL |
Each for $43.79
Case of 12 Each for $525.48
|
|
|||||
|
02992B
|
DWK Life Sciences
FS1503560 |
60 mL |
Each for $45.09
Case of 12 Each for $541.08
|
|
|||||
Description
Convenient all-in-one configuration
- Constructed of GG-33 borosilicate glass
- Ground-in glass pipet and rubber bulb included
Specifications
| Ground in Glass pipet and rubber bulb | |
| Borosilicate Glass |