Learn More
DWK Life Sciences Kimble™ Clear Glass Graduated AC Medium Rounds with PTFE-Faced LDPE Foam-lined Caps
Wide-mouth design for efficient addition and removal of contents
Supplier:  DWK Life Sciences 5810448C26
 View more versions of this product
Catalog No. 13757025
Only null left
Description
Description
  • AC Medium Round bottles are ideal for storage for solids, liquids, powders and specimens
  • Clear glass allows for easy viewing of contents
  • Choose from a variety of cap / liner combinations in Bulk Packs
  • Manufactured from soda-lime glass conforming to USP Type III and ASTM E438, Type II requirements
Specifications
Chemical Identifiers
Clear
Black
Phenolic
Glass
PTFE/LDPE
Screw Cap
Specifications
4 oz., 125 mL
4 oz.
2 in.
144/Cs.
102 mm
4.01 in.
Phenolic
51 mm
125 mL
Clear
Glass
Black
AC Medium Round Bottle
PTFE/LDPE
48-400 mm
Screw Cap
Product Suggestions
Product Suggestions