missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Microscale Organic Chemistry LABSETS™ and Kits Components
Components for microscale organic chemistry LABSETS
Supplier: DWK Life Sciences 7480131000
Description
- Microflex Kits feature special chaste connectors and specially tooled glassware components, providing great versatility
- They fit easily into lab bench drawers
Chemical Identifiers
| Microscale organic chemistry |
Specifications
| Microscale organic chemistry | |
| Component part of Williamson standard organic chemistry labsets and kits | |
| 25/Pk. | |
| Reaction Tube |