Learn More
DWK Life Sciences Kimble™ Silicone Stoppers, Single Hole
Kimble™ Silicone Stoppers are general purpose single-hole silicone rubber stoppers. They are steam autoclavable and an opaque yellowish-white in color. For use with Kimble™ Kontes™ Filtration Assemblies.
$82.60 - $107.07
| Catalog Number | Mfr. No. | Standard Taper Joint | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Standard Taper Joint | Price | Quantity | |||||
|
K953715-0501
|
DWK Life Sciences
9537150501 |
No. 5 |
Each for $82.60
|
|
|||||
|
K953715-0801
|
DWK Life Sciences
9537150801 |
Each for $107.07
|
|
||||||
|
K9537630801
|
DWK Life Sciences
9537630801 |
No. 8 |
Each for $103.33
|
|
|||||
Description
- Product Type: Stopper
- Material: Silicone Rubber
- Color: White/Yellow
- Height: 25 mm
- Autoclavable: Yes
- Use with: Kimble™ Kontes™ Filtration Assemblies