Learn More
DWK Life Sciences KIMBLE™ RAY-SORB™ Amber Heavy Duty Wide Mouth Volumetric Flask, PTFE Stopper
Stoppers are color coded for easy identification
$547.23 - $1948.90
Specifications
Closure Material PTFE
Color Red
Material Borosilicate Glass
Mouth Style Wide
Bottom Shape Flat
View More Specs
Products 11
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
13-756-711
View Documents
DWK Life Sciences
92822F5
5 mL
Case of 6 Each for $787.47
Only null left
13-756-712
View Documents
DWK Life Sciences
92822F10
10 mL
Case of 6 Each for $845.73
Only null left
13-756-713
View Documents
DWK Life Sciences
92822F20
20 mL
Case of 6 Each for $970.33
Only null left
13-756-715
View Documents
DWK Life Sciences
92822F50
50 mL
Case of 6 Each for $1,122.20
Only null left
13-756-714
View Documents
DWK Life Sciences
92822F25
25 mL
Case of 6 Each for $870.27
Only null left
13-756-716
View Documents
DWK Life Sciences
92822F100
100 mL
Case of 6 Each for $1,426.23
Only null left
13-756-717
View Documents
DWK Life Sciences
92822F200
200 mL
Case of 6 Each for $1,410.67
Only null left
13-756-720
View Documents
DWK Life Sciences
92822F1000
1000 mL
Case for $547.23
Only null left
13-756-721
View Documents
DWK Life Sciences
92822F2000
2000 mL
Case for $807.33
Only null left
13-756-718
View Documents
DWK Life Sciences
92822F250
250 mL
Case of 6 Each for $1,484.93
Only null left
13-756-719
View Documents
DWK Life Sciences
92822F500
500 mL
Case of 6 Each for $1,948.90
Only null left
Description
Description
RAY-SORB processing protects contents from harmful UV light
  • Wide mouth makes pipet access easy
  • Heavy, uniform walls provide superior strength, durability and safety
  • Large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Calibrated “To Contain”
  • Sizes from 5 to 250mL are calibrated to meet ASTM™ E288
Specifications
Specifications
PTFE
Borosilicate Glass
Flat
Red
Wide
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'