Learn More
DWK Life Sciences Kimble™ Flat Bottom Screw threaded Culture Tubes
Without caps
$921.16 - $1077.67
Specifications
Certifications/Compliance USP Type I, ASTM E 438 Type I, Class B
Material Borosilicate Glass
Disposable Single-use
For Use With For tissue culture and clinical chemistry
Label Type None
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Length (Metric) Diameter (Metric) Thread Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Length (Metric) Diameter (Metric) Thread Size Price Quantity  
11-301-235
View Documents
DWK Life Sciences
90H13100F
8 mL 100 mm 13 mm 13-415
Case of 1000 Each for $1,077.67
Only null left
11-301-233
View Documents
DWK Life Sciences
90K16100F
12 mL 100 mm 16 mm 15-415
Case of 1000 Each for $1,033.33
Only null left
14-955-402
View Documents
DWK Life Sciences
90K16125F
16 mL 125 mm 16 mm 15-415
Case of 1000 Each for $921.16
Only null left
Description
Description
Without caps
Suitable for tissue culture work and general bacteriological use
  • Borosilicate glass construction
  • Flat bottom and without a marking area
  • Trays are shrink wrapped in plastic film in modular trays
  • Supplied without caps
Specifications
Specifications
USP Type I, ASTM E 438 Type I, Class B
Single-use
None
Borosilicate Glass
For tissue culture and clinical chemistry
Shrink Wrapped Tray
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'