Learn More
DWK Life Sciences Kimble™ Flat Bottom Screw threaded Culture Tubes
Without caps
Supplier:  DWK Life Sciences 90H13100F
 View more versions of this product
Catalog No. 11-301-235
Only null left
Explore available promotions
Description
Description
Without caps
  • Borosilicate glass
  • Shrink wrap trays
Specifications
Specifications
USP Type I, ASTM E 438 Type I, Class B
100 mm
Single-use
None
13-415
8 mL
Borosilicate Glass
For tissue culture and clinical chemistry
Shrink Wrapped Tray
13 mm
Product Suggestions
Product Suggestions