Learn More
DWK Life Sciences Kimble™ Economy Columns Sample Diffusion Disc
Flex-Column™ sample diffusion discs protect the gel bed from being disrupted during sample application whenever a flow adapter is not used.
$90.87 - $114.50
Specifications
For Use With Kimble-Chase Economy Columns
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity  
K420404-1000
View Documents
DWK Life Sciences
4204041000
1 cm
Each for $90.87
Only null left
K420404-2500
View Documents
DWK Life Sciences
4204042500
2.5 cm
Each for $114.50
Only null left
K420404-1500
View Documents
DWK Life Sciences
4204041500
1.5 cm
Each for $100.67
Only null left
Description
Description
Flex-Column™ sample diffusion discs protect the gel bed from being disrupted during sample application whenever a flow adapter is not used.
Install on top of column packing to protect packing from disruption. Polyethylene disc is sealed to transparent acrylic ring.
Specifications
Specifications
Kimble-Chase Economy Columns
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'