Learn More
DWK Life Sciences Kimble™ Economy Columns Sample Diffusion Disc
Flex-Column™ sample diffusion discs protect the gel bed from being disrupted during sample application whenever a flow adapter is not used.
Supplier:  DWK Life Sciences 4204041000
 View more versions of this product
Catalog No. K420404-1000
Only null left
Explore available promotions
Description
Description
Flex-Column™ sample diffusion discs protect the gel bed from being disrupted during sample application whenever a flow adapter is not used.
  • A transparent acrylic ring is sealed to a 20um porosity HDPE disc which protects the top of the gel bed when the sample is manually applied
  • A crossbar on the top of the acrylic ring permits easy removal of the sample diffusion disc from the column.
Specifications
Specifications
Sample Diffusion Disc
1 cm
Kimble-Chase Economy Columns
Product Suggestions
Product Suggestions