DWK Life Sciences Kimble™ Economy Columns Sample Diffusion Disc
Flex-Column™ sample diffusion discs protect the gel bed from being disrupted during sample application whenever a flow adapter is not used.
Supplier: DWK Life Sciences 4204042500
Description
Flex-Column™ sample diffusion discs protect the gel bed from being disrupted during sample application whenever a flow adapter is not used.
- A transparent acrylic ring is sealed to a 20um porosity HDPE disc which protects the top of the gel bed when the sample is manually applied
- A crossbar on the top of the acrylic ring permits easy removal of the sample diffusion disc from the column.
Specifications
2.5 cm | |
Kimble-Chase Economy Columns |