missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A40 Blue Breathable Back Coveralls with Hood
Breathable coveralls with a blue back, from shoulders to ankles, feature a microporous film laminate fabric that provides barrier protection against hazardous particulates, aerosols, and low-hazard liquid splash.
$199.50 - $430.84
Specifications
| Product Type | General Purpose Coveralls |
|---|---|
| Gender | Unisex |
| Material | Microporous Film Laminate |
| Color | White, Blue |
| Disposable | Limited-use |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
19-048-600
|
Ansell
37591 |
Large |
Case of 25 Each for $199.50
|
|
|||||
|
19-048-601
|
Ansell
37163 |
X-Large |
Case of 25 Each for $430.84
|
|
|||||
|
19-048-602
|
Ansell
37164 |
2X-Large |
Case of 25 Each for $199.50
|
|
|||||
|
19-048-603
|
Ansell
37165 |
3X-Large |
Case of 25 Each for $212.49
|
|
|||||
|
19-048-604
|
Ansell
38508 |
4X-Large |
Case of 25 Each for $242.40
|
|
|||||
|
19-048-605
|
Ansell
37584 |
5X-Large/6X-Large |
Case of 25 Each for $250.26
|
|
|||||
Description
Breathable coveralls with a blue back, from shoulders to ankles, feature a microporous film laminate fabric that provides barrier protection against hazardous particulates, aerosols, and low-hazard liquid splash.
- Breathable back, made of SMS fabric, from the shoulders to ankles, which helps keep the wearer cool
- Each protective suit comes with a zipper front, a hood and elastic wrists and ankles
- Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals and dry particulates
- Material gives wearers a better liquid barrier and a better particulate barrier than Dupont Tyvek
- Pass NFPA 99 for antistatic materials
- Pass ASTM F1670 testing for penetration of blood and bodily fluids
- Meet the ANSI/ISEA 101-1996 sizing standards
- Popular industries using this product include aviation, pharmaceutical manufacturing, wind blade manufacturing, liquid handling, general manufacturing
Specifications
| General Purpose Coveralls | |
| Microporous Film Laminate | |
| Limited-use | |
| Elastic | |
| Hooded | |
| Front Zipper | |
| Non-sterile | |
| Anti-static | |
| ANSI/ISEA 101-1996, ASTM F1670, ASTM F1671, ASTM F903, NFPA 99 | |
| KleenGuard™ A40 |
| Unisex | |
| White, Blue | |
| Elastic | |
| Elastic | |
| Elastic | |
| Serged | |
| Non-toxic Particulates, Bloodborne Pathogens | |
| Breathable | |
| Non-woven Microporous Film Laminate Barrier Over SMS | |
| Healthcare, Fiberglass Manufacturing, Liquid Handling, Utilities, Pressure Washing, Tank Cleaning, Wind Blade Manufacturing, General Manufacturing |