missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A40 Blue Breathable Back Coveralls with Hood

Description
Breathable coveralls with a blue back, from shoulders to ankles, feature a microporous film laminate fabric that provides barrier protection against hazardous particulates, aerosols, and low-hazard liquid splash.
- Breathable back, made of SMS fabric, from the shoulders to ankles, which helps keep the wearer cool
- Each protective suit comes with a zipper front, a hood and elastic wrists and ankles
- Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals and dry particulates
- Material gives wearers a better liquid barrier and a better particulate barrier than Dupont Tyvek
- Pass NFPA 99 for antistatic materials
- Pass ASTM F1670 testing for penetration of blood and bodily fluids
- Meet the ANSI/ISEA 101-1996 sizing standards
- Popular industries using this product include aviation, pharmaceutical manufacturing, wind blade manufacturing, liquid handling, general manufacturing
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | Large |
| Gender | Unisex |
| Material | Microporous Film Laminate |
| Color | White, Blue |
| Disposable | Limited-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |