missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A40 Blue Breathable Back Coveralls with Hood
Breathable coveralls with a blue back, from shoulders to ankles, feature a microporous film laminate fabric that provides barrier protection against hazardous particulates, aerosols, and low-hazard liquid splash.
Supplier: Ansell 37584
Description
Breathable coveralls with a blue back, from shoulders to ankles, feature a microporous film laminate fabric that provides barrier protection against hazardous particulates, aerosols, and low-hazard liquid splash.
- Breathable back, made of SMS fabric, from the shoulders to ankles, which helps keep the wearer cool
- Each protective suit comes with a zipper front, a hood and elastic wrists and ankles
- Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals and dry particulates
- Material gives wearers a better liquid barrier and a better particulate barrier than Dupont Tyvek
- Pass NFPA 99 for antistatic materials
- Pass ASTM F1670 testing for penetration of blood and bodily fluids
- Meet the ANSI/ISEA 101-1996 sizing standards
- Popular industries using this product include aviation, pharmaceutical manufacturing, wind blade manufacturing, liquid handling, general manufacturing
Specifications
| General Purpose Coveralls | |
| Unisex | |
| White, Blue | |
| Elastic | |
| Elastic | |
| Elastic | |
| Serged | |
| Non-toxic Particulates, Bloodborne Pathogens | |
| Breathable | |
| Non-woven Microporous Film Laminate Barrier Over SMS | |
| Healthcare, Fiberglass Manufacturing, Liquid Handling, Utilities, Pressure Washing, Tank Cleaning, Wind Blade Manufacturing, General Manufacturing |
| 5X-Large/6X-Large | |
| Microporous Film Laminate | |
| Limited-use | |
| Elastic | |
| Hooded | |
| Front Zipper | |
| Non-sterile | |
| Anti-static | |
| ANSI/ISEA 101-1996, ASTM F1670, ASTM F1671, ASTM F903, NFPA 99 | |
| KleenGuard™ A40 |