missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A30 Coveralls with Elastic Back, Wrists, Ankles with Attached Hood and Boots
Limited-use coveralls designed to protect against hazardous dry particles, aerosols, and low-hazardous limited liquid splash.
$239.64 - $264.76
Specifications
| Product Type | General Purpose Coveralls |
|---|---|
| Gender | Unisex |
| Material | SMS Fabric |
| Color | White |
| Disposable | Limited-use |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
19-170-389A
|
Ansell
46122 |
Medium |
Case of 25 Each for $249.82
|
|
|||||
|
19-170-389B
|
Ansell
46123 |
Large |
Case of 25 Each for $239.64
|
|
|||||
|
19-170-389D
|
Ansell
46125 |
2X-Large |
Case of 25 Each for $239.64
|
|
|||||
|
19-170-389E
|
Ansell
46126 |
3X-Large |
Case of 21 Each for $251.40
|
|
|||||
|
19-170-389C
|
Ansell
46124 |
X-Large |
Case of 25 Each for $249.82
|
|
|||||
|
19-170-389F
|
Ansell
46127 |
4X-Large |
Case of 21 Each for $244.26
|
|
|||||
|
19-188-583
|
Ansell
27239 |
6X-Large |
Case of 21 Each for $264.76
|
|
|||||
Description
- SMS Fabric incorporates a web of microfibers that filter particulates and water-based liquids
- Suitable for many applications including pharmaceutical research, food processing and more
- Protection from non hazardous particles and liquids
- Strong and abrasion resistant; suitable for multiple wearings
- Seamless front to provide more protection in the primary exposure areas
- Zipper front with 1 in. (2.5 cm) flap and elastic back
- Elastic back, wrists and ankles and attached hood and boots
Specifications
| General Purpose Coveralls | |
| SMS Fabric | |
| Limited-use | |
| Elastic | |
| Attached, Open Face | |
| Non-sterile | |
| Elastic | |
| KleenGuard™ A30 |
| Unisex | |
| White | |
| Elastic | |
| Hooded | |
| Front Zipper | |
| Particle, Liquid | |
| NFPA 99 | |
| Pharmaceutical Research, Aerospace, Maintenance, General Manufacturing, Food |