missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A30 Coveralls with Elastic Back, Wrists, Ankles with Attached Hood and Boots
Limited-use coveralls designed to protect against hazardous dry particles, aerosols, and low-hazardous limited liquid splash.
Supplier: Ansell 46122
Description
- SMS Fabric incorporates a web of microfibers that filter particulates and water-based liquids
- Suitable for many applications including pharmaceutical research, food processing and more
- Protection from non hazardous particles and liquids
- Strong and abrasion resistant; suitable for multiple wearings
- Seamless front to provide more protection in the primary exposure areas
- Zipper front with 1 in. (2.5 cm) flap and elastic back
- Elastic back, wrists and ankles and attached hood and boots
Specifications
| General Purpose Coveralls | |
| Unisex | |
| White | |
| Elastic | |
| Hooded | |
| Front Zipper | |
| Particle, Liquid | |
| NFPA 99 | |
| Pharmaceutical Research, Aerospace, Maintenance, General Manufacturing, Food |
| Medium | |
| SMS Fabric | |
| Limited-use | |
| Elastic | |
| Attached, Open Face | |
| Non-sterile | |
| Elastic | |
| KleenGuard™ A30 |