Learn More
Ansell KleenGuard™ A30 Coveralls with Elastic Back, Wrists, Ankles with Attached Hood and Boots
Limited-use coveralls designed to protect against hazardous dry particles, aerosols, and low-hazardous limited liquid splash.
Supplier:  Ansell 27239
 View more versions of this product
Catalog No. 19-188-583
Only null left
Explore available promotions
Description
Description
  • SMS Fabric incorporates a web of microfibers that filter particulates and water-based liquids
  • Suitable for many applications including pharmaceutical research, food processing and more
  • Protection from non hazardous particles and liquids
  • Strong and abrasion resistant; suitable for multiple wearings
  • Seamless front to provide more protection in the primary exposure areas
  • Zipper front with 1 in. (2.5 cm) flap and elastic back
  • Elastic back, wrists and ankles and attached hood and boots
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Hooded
Front Zipper
Particle, Liquid
NFPA 99
Pharmaceutical Research, Aerospace, Maintenance, General Manufacturing, Food
6X-Large
SMS Fabric
Limited-use
Elastic
Attached, Open Face
Non-sterile
Elastic
KleenGuard™ A30
Product Suggestions
Product Suggestions