Learn More
Ansell Kimtech™ Pure G5 Co-Polymer Gloves
Non-sterile co-polymer gloves offer a comfortable fit and a smooth finish for tactile sensitivity. Recommended for Class 100/ISO 5 or higher cleanrooms.
Supplier:  Ansell 61001
 View more versions of this product
Catalog No. 19-048587B
Only null left
Explore available promotions
Description
Description
  • Proprietary polymer composition and processing minimize dry particle release and chemical extractables
  • ESD properties
  • Manufacturer-suggested ISO Class 5 (Class 100) or higher
  • Free of proteins and chemical accelerators
  • Comfort, dexterity, and durability without natural rubber latex
  • Smooth finish
  • Beaded cuff reduces tearing and eases donning and doffing
  • Ambidextrous, Non-sterile
Specifications
Specifications
ASTM D5250
Yes
Clear
No Texture
Yes
Powder Free
Non-sterile
0.07 mm
Kimtech™ Pure G5
30.5 cm
Class 100 (ISO 5)
2.5
Single-use
Ambidextrous
Co-polymer
Small
3 mil
61001
12 in.
Product Suggestions
Product Suggestions