missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ KingFisher Apex Accessories
KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source.
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
24075970
|
Thermo Scientific™
24075970 |
Matrix Heating Block |
N/A
|
|
|||||
24079910
|
Thermo Scientific™
24079910 |
96 PCR Head |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
24079920
|
Thermo Scientific™
24079920 |
Combi Head |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
24079930
|
Thermo Scientific™
24079930 |
96 DW Head |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
24079940
|
Thermo Scientific™
24079940 |
24 Combi Head |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
24075910
|
Thermo Scientific™
24075910 |
PCR Plate Heating Block |
N/A
|
|
|||||
24075920
|
Thermo Scientific™
24075920 |
96 KF Heating Block |
N/A
|
|
|||||
24075930
|
Thermo Scientific™
24075930 |
96 DW Heating Block |
N/A
|
|
|||||
24075940
|
Thermo Scientific™
24075940 |
24 DW Heating Block |
N/A
|
|
|||||
24075950
|
Thermo Scientific™
24075950 |
Heating Block 96 |
N/A
|
|
|||||
24075960
|
Thermo Scientific™
24075960 |
Heating Block 24 |
N/A
|
|
|||||
N21445
|
Thermo Scientific™
N21445 |
Storage Tube Adapter |
N/A
|
|
|||||
N21446
|
Thermo Scientific™
N21446 |
Plate Adapter |
N/A
|
|
|||||
N21447
|
Thermo Scientific™
N21447 |
UV Lamp (Replacement) |
N/A
|
|
|||||
Description
KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality
- User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
- Provides versatile and high-speed purification with reproducible results
- Easy to use interface with touch screen empowering the user to control all details of the application and purification process
- BindIx PC Software for added convenience
- Extensive selection of validated protocols for different type of applications
- Cloud library for quick protocol retrieval