missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher Apex Accessories

Catalog No. N21447
Encompass
Change view
Click to view available options
Product Type:
24 Combi Head
24 DW Heating Block
96 DW Head
96 DW Heating Block
96 KF Heating Block
96 PCR Head
Combi Head
Heating Block 24
Heating Block 96
Matrix Heating Block
PCR Plate Heating Block
Plate Adapter
Storage Tube Adapter
UV Lamp (Replacement)
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
N21447 UV Lamp (Replacement)
24075970 Matrix Heating Block
24079910 96 PCR Head
24079920 Combi Head
24079930 96 DW Head
24079940 24 Combi Head
24075910 PCR Plate Heating Block
24075920 96 KF Heating Block
24075930 96 DW Heating Block
24075940 24 DW Heating Block
24075950 Heating Block 96
24075960 Heating Block 24
N21445 Storage Tube Adapter
N21446 Plate Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. N21447 Supplier Thermo Scientific™ Supplier No. N21447

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source.

KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality

  • User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
  • Provides versatile and high-speed purification with reproducible results
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • BindIx PC Software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval

Specifications

Product Type UV Lamp (Replacement)
Disposable Single-use
For Use With (Equipment) KingFisher Apex
Quantity 1 Pc.
How many kinds (type or size) of magnets are there for the KingFisher Apex 96 DW Head (Cat. No. 24079930)?

Please see the Thermo Scientific KingFisher Flex User Manual, page 32. As you will see, there are four different heads. In terms of magnet force, it's a permanent magnet so the expected life is >100 years.