missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keracyanin chloride, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
$140.48 - $489.15
Specifications
| CAS | 18719-76-1 |
|---|---|
| Physical Form | Neat |
| Linear Formula | C27H31 ClO15 |
| MDL Number | MFCD00135404 |
| Synonym | Cyanidin-3-O-rutinoside chloride |
Description
Food and BeveragesSpecifications
| 18719-76-1 | |
| C27H31 ClO15 | |
| Cyanidin-3-O-rutinoside chloride | |
| 630.98 | |
| Analytical Standard |
| Neat | |
| MFCD00135404 | |
| Analytical Standard | |
| ≥98.0% (HPLC) |