missing translation for 'onlineSavingsMsg'
Learn More

Keracyanin chloride, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013399
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013399 1 mg
111013400 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013399 Supplier Merck Emd Millipore Supplier No. 364281MGF
Only null left
Food and Beverages

Specifications

CAS 18719-76-1
Physical Form Neat
Linear Formula C27H31 ClO15
MDL Number MFCD00135404
Synonym Cyanidin-3-O-rutinoside chloride
Type Analytical Standard
Quantity 1 mg
Formula Weight 630.98
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard