Learn More
Kanamycin monosulfate
Kanamycin monosulfate, CAS # 25389-94-0, is an aminoglycoside sulfate salt with antibacterial activity. | CAS: 25389-94-0 | C18H38N4O15S | 582.575 g/mol
Supplier: Thermo Scientific Chemicals J6127214
Description
Antibiotic. Inhibits translocation during protein biosynthesis.Kanamycin monosulfate acts as an antibiotic and used to treat various infections. It is involved in the translation of prokaryotic ribosomes arises due to bacterial cell death. It is used as a selective agent in molecular biology in order to isolate transformants that are expressing a kanamycin resistance gene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Kanamycin monosulfate is the sulfate salt form of kanamycin A
- It was isolated from Streptomyces kanamyceticus
- This compound inhibits translocation during protein biosynthesis. It binds to the 30S ribosomal subunit, causing misreading of the mRNA sequence
Application
- Kanamycin monosulfate shows antibacterial activity. It is effective against gram-positive and gram-negative bacteria, as well as Mycoplasma species
- This compound is also involved in translating prokaryotic ribosomes
- In molecular biology experiments, it is commonly used to select and isolate transformants expressing the kanamycin resistance gene, and plants that have the NPT II (APH3) gene incorporated
Specifications
| Kanamycin Monosulfate | |
| Powder | |
| White | |
| C18H38N4O15S | |
| 14,5281 | |
| Dissolve in water at 10mg/ml.Soluble in water. | |
| C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O | |
| 582.575 | |
| 582.58 |
| Kanamycin monosulfate | |
| 25389-94-0 | |
| 25 g | |
| MFCD00070253 | |
| Kanamycin A | |
| OOYGSFOGFJDDHP-IZQIRFRQSA-N | |
| (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid | |
| 134129479 |
Chemical Identifiers
| 25389-94-0 | |
| 582.575 | |
| OOYGSFOGFJDDHP-IZQIRFRQSA-N | |
| 134129479 | |
| C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O |
| C18H38N4O15S | |
| MFCD00070253 | |
| Kanamycin A | |
| (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid |
Safety and Handling
GHS H Statement
H334-H360-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May damage fertility or the unborn child.
May cause an allergic skin reaction.
P201-P202-P260-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H360FD
EINECSNumber : 246-933-9
RTECSNumber : NZ3225030
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only