missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin monosulfate, MP Biomedicals™

$269.18 - $906.28

Chemical Identifiers

CAS 25389-94-0
Molecular Formula C18H38N4O15S
Molecular Weight (g/mol) 582.575
InChI Key OOYGSFOGFJDDHP-IZQIRFRQSA-N
Synonym Kanamycin sulfate salt, Kanamycin A
PubChem CID 134129479
IUPAC Name (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15002905
View Documents
MP Biomedicals Inc
0215002905
5 g
Each for $269.18
Only null left
 
ICN15002925
View Documents
MP Biomedicals Inc
0215002925
25 g
Each for $906.28
Only null left
 
Description

Description

Specifications

Chemical Identifiers

25389-94-0
582.575
Kanamycin sulfate salt, Kanamycin A
(2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
C18H38N4O15S
OOYGSFOGFJDDHP-IZQIRFRQSA-N
134129479
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O

Specifications

Kanamycin Monosulfate
25389-94-0
5 g
Kanamycin sulfate salt, Kanamycin A
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
582.575
582.6
Powder
White
C18H38N4O15S
OOYGSFOGFJDDHP-IZQIRFRQSA-N
(2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
134129479
Kanamycin monosulfate
Videos
SDS
Documents

Documents