missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin monosulfate, Cell Culture Grade

Kanamycin monosulfate, CAS # 25389-94-0, is an aminoglycoside sulfate salt with antibacterial activity. | CAS: 25389-94-0 | C18H38N4O15S | 582.575 g/mol

Supplier:  Thermo Scientific Chemicals J6066809

 View more versions of this product

Catalog No. AAJ6066809


Only null left
Explore available promotions

Description

Description

Kanamycin monosulfate, is used as an aminoglycoside antibiotic. Kanamycin can be used as a selection agent in several types of isolation media.Kanamycin sulfate from Streptomyces kanamyceticus has been used in tissue culture media. Kanamycin sulfate can also be used as a selection agent for cells transformed with kanamycin B resistance gene. It is recommended for use in cell culture applications at 100mg/mL.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Kanamycin monosulfate is the sulfate salt form of kanamycin A

• It was isolated from Streptomyces kanamyceticus

• This compound inhibits translocation during protein biosynthesis. It binds to the 30S ribosomal subunit, causing misreading of the mRNA sequence

Application

• Kanamycin monosulfate shows antibacterial activity. It is effective against gram-positive and gram-negative bacteria, as well as Mycoplasma species

• This compound is also involved in translating prokaryotic ribosomes

• In molecular biology experiments, it is commonly used to select and isolate transformants expressing the kanamycin resistance gene, and plants that have the NPT II (APH3) gene incorporated

Literature References

Patel, K. N.; Limgavkar, R. S.; Raval, H. G.; Patel, K. G.; Gandhi, T. R. High-Performance Liquid Chromatographic Determination of Cefalexin Monohydrate and Kanamycin Monosulfate with Precolumn Derivatization. J. Liq. Chromatogr. Related Technol. 2015, 38 (6), 716-721.

Chi, Z.; Rong, Y. J.; Li, Y.; Tang, M. J.; Chi, Z. M. Biosurfactins production by Bacillus amyloliquefaciens R3 and their antibacterial activity against multi-drug resistant pathogenic E. coli. Bioprocess. Biosyst. Eng. 2015, 38 (5), 853-861.

Specifications

Specifications

Kanamycin Monosulfate
Powder
White
C18H38N4O15S
14,5281
Soluble in water at 10mg/ml
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
582.575
582.58
Kanamycin monosulfate
25389-94-0
10 g
MFCD00070253
Kanamycin A
OOYGSFOGFJDDHP-IZQIRFRQSA-N
(2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
134129479
Cell Culture

Chemical Identifiers

25389-94-0
582.575
OOYGSFOGFJDDHP-IZQIRFRQSA-N
134129479
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
C18H38N4O15S
MFCD00070253
Kanamycin A
(2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H360-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May damage fertility or the unborn child.
May cause an allergic skin reaction.

P201-P202-P260-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H360FD

EINECSNumber : 246-933-9

RTECSNumber : NZ3225030

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only