missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ JW 480

Catalog No. 432710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
432710 10 mg
432750 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 432710 Supplier Tocris Bioscience™ Supplier No. 4327/10
Only null left

Potent and selective inhibitor of serine hydrolase KIAA1363 (AADACL1)

Potent and selective inhibitor of the serine hydrolase enzyme KIAA1363 (AADACL1) (IC50 = 20 nM in mouse brain). Impairs migration, invasion, survival and tumor growth of prostate cancer cell lines in vivo. Reduces monoalkylglycerol ether (MAGE) levels.

Chemical Identifiers

CAS 1354359-53-7
Molecular Formula C22H23NO2

Specifications

Quantity 10 mg
Formula Weight 333.42
Chemical Name or Material JW 480
Recommended Storage Store at -20°C