Learn More
United Scientific™ Jupiter Electronic Pipette, 8-Channel
Versatile multi-channel pipettes deliver fast and accurate results with three operating modes: pipetting, step dispensing, and diluting.
Supplier:  United Scientific™ UNEMCH0100
 View more versions of this product
Catalog No. S00057
Only null left
Explore available promotions
Alert:
Prop 65
Description
Description
  • Increase efficiency and speed with the Jupiter 8-Channel Electronic Pipettes designed for precise, flexible liquid handling.
  • Supports multiple functions including pipetting, repetitive dispensing, and diluting.
  • Simple two-knob interface and clear digital display allow quick switching between modes, volumes, and speeds.
  • High-performance stepper motor ensures accuracy, repeatability, and adjustable aspiration/dispense speeds.
  • Lightweight and well-balanced pipette reduces fatigue during extended use; compatible with most universal tip brands.
  • Intuitive LCD interface and 360° rotating tip ejector collar for smooth, comfortable handling.
  • Long-lasting lithium-ion battery recharges via AC adapter or USB; includes wall-mounted charging stand.
  • Calibrated and tested in compliance with ISO8655-6 / DIN12650 standards.
  • Available in multiple volume capacities.
Specifications
Specifications
Lithium Ion
100, 200, 300, 350 μL
Not Autoclavable
Reusable
Ergonomic
Jupiter
No
Multichannel Pipette with Variable Volume Setting
Digital
Blue
ISO8655-6 / DIN12650
No
White
Pipette Tip 100, 200, 300, 350 μL
Plastic
8
Non-sterile
10 to 100 μL
0.1 μL
Product Suggestions
Product Suggestions