Learn More
United Scientific™ Jupiter Electronic Pipette, 8-Channel
Versatile multi-channel pipettes deliver fast and accurate results with three operating modes: pipetting, step dispensing, and diluting.
$3045.63 - $3045.63
Specifications
Battery Type Lithium Ion
Certifications/Compliance ISO8655-6 / DIN12650
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color White
View More Specs
Alert:
Prop 65
Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
S00056
View Documents
United Scientific™
UNEMCH0010
0.5 to 10 μL
Each for $3,045.63
Only null left
S00057
View Documents
United Scientific™
UNEMCH0100
10 to 100 μL
Each for $3,045.63
Only null left
S00058
View Documents
United Scientific™
UNEMCH0300
15 to 300 μL
Each for $3,045.63
Only null left
Description
Description
  • Increase efficiency and speed with the Jupiter 8-Channel Electronic Pipettes designed for precise, flexible liquid handling.
  • Supports multiple functions including pipetting, repetitive dispensing, and diluting.
  • Simple two-knob interface and clear digital display allow quick switching between modes, volumes, and speeds.
  • High-performance stepper motor ensures accuracy, repeatability, and adjustable aspiration/dispense speeds.
  • Lightweight and well-balanced pipette reduces fatigue during extended use; compatible with most universal tip brands.
  • Intuitive LCD interface and 360° rotating tip ejector collar for smooth, comfortable handling.
  • Long-lasting lithium-ion battery recharges via AC adapter or USB; includes wall-mounted charging stand.
  • Calibrated and tested in compliance with ISO8655-6 / DIN12650 standards.
  • Available in multiple volume capacities.
Specifications
Specifications
Lithium Ion
No
White
Ergonomic
Jupiter
No
Multichannel Pipette with Variable Volume Setting
Blue
ISO8655-6 / DIN12650
Not Autoclavable
Reusable
Plastic
8
Non-sterile
Digital
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'