missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JT010
Description
JT010 is a potent and selective TRPA1 channel activator (EC50 = 0.65 nM); activates the TRPA1 channel by covalently and site-selectively binding to Cys621. Exhibits selectivity for TRPA1 over TRPV1, TRPV3, TRPV4, TRPM2, TRPM8, and TRPC5 channels (EC50 values are >1 μM).
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-N-[4-(4-methoxyphenyl)-2-thiazolyl]-N-(3-methoxypropyl)acetamide |
| CAS | 917562-33-5 |
| Quantity | 5 mg |
| Molecular Formula | C16H19ClN2O3S |
| Formula Weight | 354.85 |
| Purity | 98% |