missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JNJ 63533054
Description
JNJ 63533054 is a potent and selective GPR139 agonist (EC50 = 16 nM). Selective for GPR139 over a panel of GPCRs, ion channels and transporters, including GPR142. Brain and cell penetrant. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-N-[2-oxo-2-[[(1S)-1-phenylethyl]amino]ethyl]benzamide |
| CAS | 1802326-66-4 |
| Quantity | 50 mg |
| Target | Orphan 7-TM Receptor Agonists |
| Molecular Formula | C17H17ClN2O2 |
| Formula Weight | 316.78 |
| Purity | >98% |