missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JBJ-03-142-02
Description
JBJ-03-142-02 is a highly potent epidermal growth factor receptor (EGFR) and ErbB2 (human epidermal growth factor receptor; HER2) inhibitor. Inhibits wild type and tumorgenic mutated receptors (exon 20 insertion; Ex20Ins). Antiproliferative in vitro (IC50 values are 7.6, 30 - 95, 3.8 and 19 - 31 nM for Ba/F3 cells expressing wild type EGFR, EGFR with Ex20Ins mutation, wild type HER2 and HER2 with Ex20Ins mutation, respectively). Also inhibits other common EGFR mutants. Antiproliferative in patient-derived lung cancer cell lines.
Specifications
Specifications
| Chemical Name or Material | (4-Fluorophenyl)methyl N-[2-[(1-methyl-1 H-pyrazol-4-yl)amino]-4-pyrimidinyl]-N-[3-[(1-oxo-2-propen-1-yl)amino]phenyl]carbamate |
| CAS | 2068806-31-3 |
| Quantity | 10 mg |
| Target | EGF Receptor Inhibitors |
| Molecular Formula | C25H22FN7O3 |
| Formula Weight | 2323 |
| Purity | 98% |