missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Janelia Fluor 646, SE
Description
Key Information: Janelia Fluor™ 646, SE is a red fluorogenic fluorescent dye; supplied with an NHS ester reactive group for the labeling of primary amines. Suitable for live cell imaging.Application: Suitable for flow cytometry, confocal microscopy, super resolution microscopy (SRM) techniques including dSTORM (in both live and fixed cells) and STED. Cell permeable.Properties and Photophysical Data: NHS ester can be converted to relevant substrate for use in self-labeling tag systems, e.g. HaloTag™ and SNAP-tag™. Can be multiplexed for two color imaging with Janelia Fluor™ 549 SE (Cat. No. 6147). Excitation and emission maxima (λ) are 646 nm and 664 nm, respectively; quantum yield = 0.54; extinction coefficient = 152,000 M-1cm-1 (measured in TFE plus 0.1% TFA); A280 correction factor = 0.19; lactone-zwitterion equilibrium constant (KL-Z) = 0.0012. Please see the protocol for further information on protein/antibody labeling and conjugation. We recommend that stock solutions of this dye are prepared in anhydrous DMF. To measure the absorbance spectrum of this dye we recommend the following solvent: ethanol or trifluoroethanol plus 0.1% TFA.We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company R&D Systems. HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc.
Specifications
Specifications
| Chemical Name or Material | 1-[7-(1-Azetidinyl)-10-[2-carboxy-5-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]phenyl]-9,9-dimethyl-9-silaanthracen-2(9H)-ylidene]azetidinium, inner salt |
| CAS | 1811539-59-9 |
| Quantity | 1 mg |
| Product Line | Tocris |
| Target | Amine-Reactive Janelia Fluor Dyes |
| Molecular Formula | C33H31N3O6Si |
| Purity | 95% |