missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ivabradine hydrochloride

Catalog No. AC466280010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466280010 1 g
AC466282500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466280010 Supplier Thermo Scientific Chemicals Supplier No. 466280010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 148849-67-6
Molecular Formula C27H37ClN2O5

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: .. Other solubilities: soluble in dmso and methanol
Melting Point 140.0°C
Formula Weight 505.05
InChI Key HLUKNZUABFFNQS-ZMBIFBSDSA-N
SMILES [H+].[Cl-].COC1=CC2=C(CC(=O)N(CCCN(C)C[C@H]3CC4=CC(OC)=C(OC)C=C34)CC2)C=C1OC
IUPAC Name hydrogen 3-[3-({[(7S)-3,4-dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl}(methyl)amino)propyl]-7,8-dimethoxy-2,3,4,5-tetrahydro-1H-3-benzazepin-2-one chloride
Molecular Weight (g/mol) 505.05
Chemical Name or Material Ivabradine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Very toxic to aquatic life.

Precautionary Statement GHS P Statement Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only