missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ivabradine hydrochloride

Catalog No. 654250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
654250 50 mg
654210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 654250 Supplier Tocris Bioscience™ Supplier No. 6542/50

HCN channel blocker; inhibits If pacemaker current

Ivabradine hydrochloride is a HCN channel blocker (IC50 is approximately 0.5 - 2.5 μM). Bradycardic agent that inhibits If pacemaker current in sinoatrial node cells.

Specifications

Chemical Name or Material 3-[3-[[[(7S)-3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylamino]propyl]-1,3,4,5-tetrahydro-7,8-dimethoxy-2H-3-benzazepin-2-one hydrochloride
CAS 148849-67-6
Quantity 50 mg
Molecular Formula C27H36N2O5.HCl
Purity 98%