missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Itaconic Acid 99.0+%, TCI America™
Chemical Identifiers
| CAS | 97-65-4 |
|---|---|
| Molecular Formula | C5H6O4 |
| Molecular Weight (g/mol) | 130.099 |
| MDL Number | MFCD00004260 |
| InChI Key | LVHBHZANLOWSRM-UHFFFAOYSA-N |
| Synonym | itaconic acid, 2-methylenesuccinic acid, methylenesuccinic acid, methylenebutanedioic acid, propylenedicarboxylic acid, itaconate, 2-propene-1,2-dicarboxylic acid, butanedioic acid, methylene, succinic acid, methylene, 2-methylenebutanedioic acid |
| PubChem CID | 811 |
| ChEBI | CHEBI:30838 |
| IUPAC Name | 2-methylidenebutanedioic acid |
| SMILES | C=C(CC(=O)O)C(=O)O |
Chemical Identifiers
| C5H6O4 | |
| MFCD00004260 | |
| itaconic acid, 2-methylenesuccinic acid, methylenesuccinic acid, methylenebutanedioic acid, propylenedicarboxylic acid, itaconate, 2-propene-1,2-dicarboxylic acid, butanedioic acid, methylene, succinic acid, methylene, 2-methylenebutanedioic acid | |
| CHEBI:30838 | |
| C=C(CC(=O)O)C(=O)O |
Specifications
| 97-65-4 | |
| White | |
| MFCD00004260 | |
| LVHBHZANLOWSRM-UHFFFAOYSA-N | |
| 2-methylidenebutanedioic acid | |
| 811 | |
| 130.10 | |
| Crystalline Powder |
| 167°C | |
| C5H6O4 | |
| itaconic acid, 2-methylenesuccinic acid, methylenesuccinic acid, methylenebutanedioic acid, propylenedicarboxylic acid, itaconate, 2-propene-1,2-dicarboxylic acid, butanedioic acid, methylene, succinic acid, methylene, 2-methylenebutanedioic acid | |
| C=C(CC(=O)O)C(=O)O | |
| 130.099 | |
| CHEBI:30838 | |
| ≥99.0% (T) | |
| Itaconic Acid |
Safety and Handling
EINECSNumber : (2)-1125
TSCA : Yes