missing translation for 'onlineSavingsMsg'
Learn More

Itaconic Acid 99.0+%, TCI America™

Catalog No. M022325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M022325G 25 g
M0223500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M022325G Supplier TCI America Supplier No. M022325G

Chemical Identifiers

CAS 97-65-4
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.099
MDL Number MFCD00004260
InChI Key LVHBHZANLOWSRM-UHFFFAOYSA-N
Synonym itaconic acid, 2-methylenesuccinic acid, methylenesuccinic acid, methylenebutanedioic acid, propylenedicarboxylic acid, itaconate, 2-propene-1,2-dicarboxylic acid, butanedioic acid, methylene, succinic acid, methylene, 2-methylenebutanedioic acid
PubChem CID 811
ChEBI CHEBI:30838
IUPAC Name 2-methylidenebutanedioic acid
SMILES C=C(CC(=O)O)C(=O)O

Specifications

Melting Point 167°C
Color White
Quantity 25 g
Formula Weight 130.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Itaconic Acid
EINECSNumber (2)-1125
TSCA Yes