missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isovanillic Acid 98.0+%, TCI America™
$95.06 - $300.11
Chemical Identifiers
| CAS | 645-08-9 |
|---|---|
| Molecular Formula | C8H8O4 |
| Molecular Weight (g/mol) | 168.148 |
| MDL Number | MFCD00002507 |
| InChI Key | LBKFGYZQBSGRHY-UHFFFAOYSA-N |
| Synonym | isovanillic acid, acide isovanillique, 3-hydroxyanisic acid, 3-hydroxy-p-anisic acid, benzoic acid, 3-hydroxy-4-methoxy, p-anisic acid, 3-hydroxy, chembl88700, 3-hydroxy-4-methoxy-benzoic acid, 3-hydroxy-4-methoxybenzoicacid, 5-carboxyguaiacol |
| PubChem CID | 12575 |
| ChEBI | CHEBI:63798 |
| IUPAC Name | 3-hydroxy-4-methoxybenzoic acid |
| SMILES | COC1=C(C=C(C=C1)C(=O)O)O |
Chemical Identifiers
| 645-08-9 | |
| 168.148 | |
| LBKFGYZQBSGRHY-UHFFFAOYSA-N | |
| 12575 | |
| 3-hydroxy-4-methoxybenzoic acid |
| C8H8O4 | |
| MFCD00002507 | |
| isovanillic acid, acide isovanillique, 3-hydroxyanisic acid, 3-hydroxy-p-anisic acid, benzoic acid, 3-hydroxy-4-methoxy, p-anisic acid, 3-hydroxy, chembl88700, 3-hydroxy-4-methoxy-benzoic acid, 3-hydroxy-4-methoxybenzoicacid, 5-carboxyguaiacol | |
| CHEBI:63798 | |
| COC1=C(C=C(C=C1)C(=O)O)O |
Specifications
| 645-08-9 | |
| C8H8O4 | |
| 5 g | |
| LBKFGYZQBSGRHY-UHFFFAOYSA-N | |
| 3-hydroxy-4-methoxybenzoic acid | |
| 12575 | |
| 168.15 | |
| Crystalline Powder |
| 253°C | |
| MFCD00002507 | |
| isovanillic acid, acide isovanillique, 3-hydroxyanisic acid, 3-hydroxy-p-anisic acid, benzoic acid, 3-hydroxy-4-methoxy, p-anisic acid, 3-hydroxy, chembl88700, 3-hydroxy-4-methoxy-benzoic acid, 3-hydroxy-4-methoxybenzoicacid, 5-carboxyguaiacol | |
| COC1=C(C=C(C=C1)C(=O)O)O | |
| 168.148 | |
| CHEBI:63798 | |
| ≥98.0% (HPLC,T) | |
| Isovanillic Acid |
Safety and Handling
RTECSNumber : BZ4850000
TSCA : No