missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isovanillic Acid 98.0+%, TCI America™
Supplier: TCI America I029525G
Specifications
| Isovanillic Acid | |
| 253°C | |
| MFCD00002507 | |
| isovanillic acid, acide isovanillique, 3-hydroxyanisic acid, 3-hydroxy-p-anisic acid, benzoic acid, 3-hydroxy-4-methoxy, p-anisic acid, 3-hydroxy, chembl88700, 3-hydroxy-4-methoxy-benzoic acid, 3-hydroxy-4-methoxybenzoicacid, 5-carboxyguaiacol | |
| COC1=C(C=C(C=C1)C(=O)O)O | |
| 168.148 | |
| CHEBI:63798 | |
| ≥98.0% (HPLC,T) |
| 645-08-9 | |
| C8H8O4 | |
| 25 g | |
| LBKFGYZQBSGRHY-UHFFFAOYSA-N | |
| 3-hydroxy-4-methoxybenzoic acid | |
| 12575 | |
| 168.15 | |
| Crystalline Powder |
Chemical Identifiers
| 645-08-9 | |
| 168.148 | |
| LBKFGYZQBSGRHY-UHFFFAOYSA-N | |
| 12575 | |
| 3-hydroxy-4-methoxybenzoic acid |
| C8H8O4 | |
| MFCD00002507 | |
| isovanillic acid, acide isovanillique, 3-hydroxyanisic acid, 3-hydroxy-p-anisic acid, benzoic acid, 3-hydroxy-4-methoxy, p-anisic acid, 3-hydroxy, chembl88700, 3-hydroxy-4-methoxy-benzoic acid, 3-hydroxy-4-methoxybenzoicacid, 5-carboxyguaiacol | |
| CHEBI:63798 | |
| COC1=C(C=C(C=C1)C(=O)O)O |
Safety and Handling
RTECSNumber : BZ4850000
TSCA : No