missing translation for 'onlineSavingsMsg'
Learn More

Isoproterenol Hydrochloride 99.0+%, TCI America™

Catalog No. p-7131298
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02605G 5 g
I026025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I02605G Supplier TCI America Supplier No. I02605G
Only null left

Chemical Identifiers

CAS 51-30-9
Molecular Formula C11H18ClNO3
Molecular Weight (g/mol) 247.72
MDL Number MFCD00012603,MFCD00064548
InChI Key IROWCYIEJAOFOW-UHFFFAOYNA-N
Synonym isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso
PubChem CID 5807
IUPAC Name hydrogen 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol chloride
SMILES [H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1

Specifications

Color White
Quantity 5 g
Formula Weight 247.72
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Isoproterenol Hydrochloride
RTECSNumber DO1925000
TSCA No