accessibility menu, dialog, popup

UserName

CAS RN 51-30-9

IUPAC Name: hydrogen 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol chloride
Synonyms: hydrogen isoproterenol chloride
Molecular Weight (g/mol): 247.72
Molecular Formula: C11H18ClNO3
InChi Key: IROWCYIEJAOFOW-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)