missing translation for 'onlineSavingsMsg'
Learn More

Isopropyltriphenylphosphonium Iodide 98.0+%, TCI America™

Catalog No. I05525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05525G 5 g
I055225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05525G Supplier TCI America Supplier No. I05525G

Chemical Identifiers

CAS 24470-78-8
Molecular Formula C21H22IP
Molecular Weight (g/mol) 432.29
MDL Number MFCD00031595
InChI Key HHBXWXJLQYJJBW-UHFFFAOYSA-M
Synonym isopropyltriphenylphosphonium iodide, phosphonium, 1-methylethyl triphenyl-, iodide, isopropyltriphenylphosphanium iodide, isopropyl triphenyl phosphonium iodide, triphenyl propan-2-yl phosphanium iodide, i-propyl triphenylphosphonium iodide, isopropyl triphenylphosphonium iodide, acmc-209gck, ksc493k7j, isopropyltriphenylphosphine iodide
PubChem CID 2723783
IUPAC Name triphenyl(propan-2-yl)phosphanium iodide
SMILES [I-].CC(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 199°C
Quantity 5 g
Formula Weight 432.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isopropyltriphenylphosphonium Iodide