accessibility menu, dialog, popup

UserName

IH3CCH3P

CAS RN 24470-78-8

IUPAC Name: triphenyl(propan-2-yl)phosphanium iodide
Synonyms: isopropyltriphenylphosphanium iodide
Molecular Weight (g/mol): 432.29
Molecular Formula: C21H22IP
InChi Key: HHBXWXJLQYJJBW-UHFFFAOYSA-M
SMILES: [I-].CC(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)