missing translation for 'onlineSavingsMsg'
Learn More

Isopropylmagnesium chloride - Lithium chloride complex, 1.3M solution in THF, AcroSeal™

Catalog No. p-3523470
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC386281000 100 mL AcroSeal™ Glass bottle
AC386288000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC386281000 Supplier Thermo Scientific Chemicals Supplier No. 386281000
Only null left

CAS: 745038-86-2 | C3H7Cl2LiMg | 145.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 745038-86-2 , 109-99-9
Molecular Formula C3H7Cl2LiMg
Molecular Weight (g/mol) 145.23
MDL Number MFCD07784514
InChI Key CWTUREABAILGIK-UHFFFAOYSA-L
Synonym turbo grignard, iprmgcl licl, i-prmgcl licl, i-prmgcl.licl, isopropylmagnesiumchloride licl, isopropylmagnesium chloride licl, isopropyl magnesium chloride licl, cwtureabailgik-uhfffaoysa-l, isopropylmagnesium lithium chloride, isopropyl magnesium chloride li-cl
PubChem CID 11275082
SMILES [Li+].[Cl-].CC(C)[Mg]Cl

Specifications

Color Gray to Yellow
Concentration 1.1 to 1.5M
Density 0.9800g/mL
Solubility Information Solubility in water: reacts.
IUPAC Name lithium;magnesium;propane;dichloride
Formula Weight 145.24
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.98
Quantity 100 mL
Flash Point −21°C
Physical Form Solution
Chemical Name or Material Isopropylmagnesium chloride - Lithium chloride complex, 14% solution in THF
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage. Highly flammable liquid and vapour. In contact with water releases flammable gases which may ignite spontaneously. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection.
Recommended Storage Product may develop some turbidity or precipitate

RUO – Research Use Only