missing translation for 'onlineSavingsMsg'
Learn More

Isopropylamine Hydrobromide 97.0+%, TCI America™

Supplier:  TCI America I10415G

 View more versions of this product

Catalog No. I10415G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Isopropylamine Hydrobromide
White
MFCD22419143
Isopropylammonium Bromide
Br.CC(C)N
140.02
140.02
Crystalline Powder
29552-58-7
C3H10BrN
5 g
WGWKNMLSVLOQJB-UHFFFAOYSA-N
propan-2-amine hydrobromide
22495069
≥97.0% (N,T)

Chemical Identifiers

29552-58-7
140.02
WGWKNMLSVLOQJB-UHFFFAOYSA-N
22495069
Br.CC(C)N
C3H10BrN
MFCD22419143
Isopropylammonium Bromide
propan-2-amine hydrobromide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions