missing translation for 'onlineSavingsMsg'
Learn More

Isopropylamine Hydrobromide 97.0+%, TCI America™

Catalog No. p-7131725
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10411G 1 g
I10415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10411G Supplier TCI America Supplier No. I10411G
Only null left

Chemical Identifiers

CAS 29552-58-7
Molecular Formula C3H10BrN
Molecular Weight (g/mol) 140.02
MDL Number MFCD22419143
InChI Key WGWKNMLSVLOQJB-UHFFFAOYSA-N
Synonym Isopropylammonium Bromide
PubChem CID 22495069
IUPAC Name propan-2-amine hydrobromide
SMILES Br.CC(C)N

Specifications

Color White
Quantity 1 g
Formula Weight 140.02
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Isopropylamine Hydrobromide