accessibility menu, dialog, popup

UserName

CH3CH3H2NHBr

CAS RN 29552-58-7

IUPAC Name: propan-2-amine hydrobromide
Synonyms: isopropylamine hydrobromide
Molecular Weight (g/mol): 140.02
Molecular Formula: C3H10BrN
InChi Key: WGWKNMLSVLOQJB-UHFFFAOYSA-N
SMILES: Br.CC(C)N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results