missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl salicylate, 99%
CAS: 607-85-2 | C10H12O3 | 180.203 g/mol
$128.15 - $361.89
Chemical Identifiers
| CAS | 607-85-2 |
|---|---|
| Molecular Formula | C10H12O3 |
| Molecular Weight (g/mol) | 180.203 |
| MDL Number | MFCD00035703 |
| InChI Key | YEULQIJMIOWCHB-UHFFFAOYSA-N |
| Synonym | isopropyl salicylate, isopropyl 2-hydroxybenzoate, salicylic acid, isopropyl ester, iso-propyl salicylate, salicylic acid isopropyl ester, isopropyl o-hydroxybenzoate, benzoic acid, 2-hydroxy-, 1-methylethyl ester, 1-methylethyl 2-hydroxybenzoate, methylethyl 2-hydroxybenzoate, opropyl salicylate |
| PubChem CID | 11838 |
| ChEBI | CHEBI:38703 |
| IUPAC Name | propan-2-yl 2-hydroxybenzoate |
| SMILES | CC(C)OC(=O)C1=CC=CC=C1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2447114
|
Thermo Scientific Chemicals
B2447114 |
25 g |
Each for $128.15
|
|
|||||
|
AAB2447122
|
Thermo Scientific Chemicals
B2447122 |
100 g |
Each for $361.89
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 607-85-2 | |
| 180.203 | |
| YEULQIJMIOWCHB-UHFFFAOYSA-N | |
| 11838 | |
| propan-2-yl 2-hydroxybenzoate |
| C10H12O3 | |
| MFCD00035703 | |
| isopropyl salicylate, isopropyl 2-hydroxybenzoate, salicylic acid, isopropyl ester, iso-propyl salicylate, salicylic acid isopropyl ester, isopropyl o-hydroxybenzoate, benzoic acid, 2-hydroxy-, 1-methylethyl ester, 1-methylethyl 2-hydroxybenzoate, methylethyl 2-hydroxybenzoate, opropyl salicylate | |
| CHEBI:38703 | |
| CC(C)OC(=O)C1=CC=CC=C1O |
Specifications
| 607-85-2 | |
| 122°C (18 mmHg) | |
| C10H12O3 | |
| 25 g | |
| YEULQIJMIOWCHB-UHFFFAOYSA-N | |
| propan-2-yl 2-hydroxybenzoate | |
| 11838 | |
| 180.2 | |
| Isopropyl salicylate |
| 1.09 | |
| >93°C (199°F) | |
| MFCD00035703 | |
| isopropyl salicylate, isopropyl 2-hydroxybenzoate, salicylic acid, isopropyl ester, iso-propyl salicylate, salicylic acid isopropyl ester, isopropyl o-hydroxybenzoate, benzoic acid, 2-hydroxy-, 1-methylethyl ester, 1-methylethyl 2-hydroxybenzoate, methylethyl 2-hydroxybenzoate, opropyl salicylate | |
| CC(C)OC(=O)C1=CC=CC=C1O | |
| 180.203 | |
| CHEBI:38703 | |
| 99% |
Safety and Handling
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 210-143-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only