accessibility menu, dialog, popup

UserName

CAS RN 607-85-2

IUPAC Name: propan-2-yl 2-hydroxybenzoate
Synonyms: isopropyl salicylate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: YEULQIJMIOWCHB-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)C1=CC=CC=C1O
Boiling Point 
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)