missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Rubbing Alcohol, USP, 68-72%, Spectrum™ Chemical

Catalog No. 18603565
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Cubitainer
Poly Bottle
Poly Drum
Steel Drum
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603565 6 x 500 mL Amber Glass Bottle
18603564 500 mL Amber Glass Bottle
18603566 1 L Amber Glass Bottle
18603567 4 L Amber Glass Bottle
18603568 4 L Poly Bottle
18603569 6 x 1 L Amber Glass Bottle
18603570 4 x 4 L Amber Glass Bottle
18603571 4 x 4 L Poly Bottle
18603573 20 L Cubitainer
18603572 20 L Poly Drum
18603574 200 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18603565 Supplier Spectrum Chemical Mfg Cor Supplier No. IS120500MLGLCS6
Only null left

IS120, 67-63-0, C3H8O

Spectrum™ Chemical Isopropyl Rubbing Alcohol, 70 Percent, USP , is a colorless and flammable chemical compound also known as Rubbing Alcohol. It is a cooling and soothing liquid for external applications to minimize the risk of infection and is not suitable for internal consumption. Spectrum Chemical USP products, including all USP alcohol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-63-0 , 7732-18-5
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 70%, 30%
Quantity 6 x 500 mL
Packaging Amber Glass Bottle
Specific Gravity 0.872-0.883
UN Number UN1219
Concentration 70%
Boiling Point 78.3°C
Percent Purity 68 to 72%
Grade USP
Chemical Name or Material Isopropyl Rubbing Alcohol